SYNB135121
Synthware™ Desiccator with lid and ceramic plate
bottle O.D. 210 mm, clear borosilicate glass 3.3
Synonym(s):
Synthware B135121, desiccator with Top Knob
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
clear borosilicate glass 3.3
packaging
pack of 1 ea
manufacturer/tradename
Kemtech B135121
bottle O.D.
210 mm
Looking for similar products? Visit Product Comparison Guide
General description
Desiccator, Clear, Id 210mm, Includes Lid & Ceramic Plate
Application
kemetch; synthware; desiccator
Features and Benefits
Organic Chemistry
Legal Information
Synthware is a trademark of Kemtech America Inc.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service