Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP6595

Sigma-Aldrich

IL-22 human

recombinant, expressed in HEK 293 cells (Ala 34 - Ile 179), ≥95% (SDS-PAGE)

Synonym(s):

IL-TIF, ZCYTO18

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

biological source

human

recombinant

expressed in HEK 293 cells (Ala 34 - Ile 179)

tag

His tagged

Assay

≥95% (SDS-PAGE)

form

lyophilized powder

mol wt

calculated mol wt 17.6 kDa (a polyhistidine tag at the N-terminus.)
observed mol wt 21-33 kDa (DTT-reduced protein.Protein migrates as 21-33 kDa in a SDS-PAGE due to glycosylation.
)

packaging

pkg of 10 μg

impurities

<1 EU/μg (LAL test)

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... IL22(50616)

General description

Interleukin-22 (IL22) is also known as cytokine Zcyto18, IL-10-related T-cell-derived-inducible factor (IL-TIF), which is belongs to the IL-10 family or IL-10 superfamily (including IL-19, IL-20, IL-24, and IL-26), a class of potent mediators of cellular inflammatory responses. IL-22 is produced by activated DC and T cells and initiates innate immune responses against bacterial pathogens especially in epithelial cells such as respiratory and gut epithelial cells. IL-22 biological activity is initiated by binding to a cell-surface complex composed of IL-22R1 and IL-10R2 receptor chains and further regulated by interactions with a soluble binding protein IL-22BP. IL-22 also promotes hepatocyte survival in the liver and epithelial cells in the lung and gut similar to IL-10.

Physical form

Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Generally Mannitol or trehalose is added as a protectant before lyophilization.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in sterile deionized water to a concentration of 100 μg/mL. Do not vortex. This solution can be stored at 2-8°C for up to 1 month. For extended storage, it is recommended to store at -20°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service