Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP6590

Sigma-Aldrich

Core Histones human

≥95% (SDS-PAGE)

Synonym(s):

HeLa Core Histones

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

Assay

≥95% (SDS-PAGE)

form

liquid

packaging

pkg of 50 and 100 μg

concentration

1 mg/mL

technique(s)

cell based assay: suitable

shipped in

dry ice

storage temp.

−70°C

General description

A nucleosome is the basic repeating unit of chromatin in which 146 base pairs of DNA wrap twice around a histone octamer consisting of two copies of each of the core histones, H2A, H2B, H3, and H4. The combination of two H2A/H2B dimers and one H3/H4 tetramer create the nucleosome core. Further, histone H1 combines with these histones to form 30nm fibers. These fibers have six nucleosomes per turn in a solenoid arrangement. Histone H3 undergoes many modifications which include acetylation, methylation, and phosphorylation that are important for regulation of gene transcription.[1]

Physical form

50 mM sodium phosphate, pH 6.9, containing 10 mM sodium chloride and 1 mM β-mercaptoethanol.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Transcription regulation by histone methylation: interplay between different covalent modifications of the core histone tails.
Zhang Y and Reinberg D
Genes & Development, 15(18), 2343-2360 (2001)
Roberto Fabian et al.
Molecules (Basel, Switzerland), 26(16) (2021-08-28)
We report data from single molecule studies on the interaction between single DNA molecules and core histones using custom-designed horizontal magnetic tweezers. The DNA-core histone complexes were formed using λ-DNA tethers, core histones, and NAP1 and were exposed to forces

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service