Skip to Content
MilliporeSigma
All Photos(1)

Documents

SRP6577

Sigma-Aldrich

TAF1 (1371-1496) GST tag human

recombinant, expressed in E. coli, ≥95% (SDS-PAGE)

Synonym(s):

BA2R, CCG1, CCGS, DYT3, KAT4, NSCL2, TAF1 RNA Polymerase II, TAF2A, TAFII250, TATA Box Binding Protein (TBP)-Associated Factor, XDP

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

Assay

≥95% (SDS-PAGE)

form

liquid

mol wt

41.7 kDa (1371-1496 aa + NT GST Tag)

packaging

pkg of 100 μg
pkg of 20 μg

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... TAF1(6872)

General description

The acetylation of histone lysine residues plays a crucial role in the epigenetic regulation of gene transcription. A bromodomain is a protein domain that recognizes acetylated lysine residues such as those on the N-terminal tails of histones. This recognition is often a prerequisite for protein-histone association and chromatin remodeling. These domains function in the linking of protein complexes to acetylated nucleosomes, thereby controlling chromatin structure and gene expression. Thus, bromodomains serve as “readers” of histone acetylation marks regulating the transcription of target promoters. TAF1 is a component of transcription factor IID, and binds to core promoter sequences at the transcription start site. TAF1 helps control transcription by both its kinase and histone acetyltransferase enzymatic activities. It interacts with transcriptional activators such as the androgen receptor to promote transcription. Disruption of TAF1 causes dystonia 3 (X-linked torsion dystonia) and decreased apoptosis. This protein product contains the first bromodomain of TAF1.

Physical form

50 mM Tris, pH 7.5, containing 500 mM sodium chloride, 5 mM β-mercaptoethanol and 5% glycerol.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service