Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP6553

Sigma-Aldrich

CIRBP/Cold-inducible RNA-binding protein human

recombinant, expressed in E. coli, ≥90% (SDS-PAGE)

Synonym(s):

A18HNRNP, CIRP

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

Assay

≥90% (SDS-PAGE)

form

lyophilized

mol wt

20.5 kDa

packaging

pkg of 1 mg
pkg of 20 μg

impurities

≤1 EU/μg endotoxin (LAL test)

UniProt accession no.

shipped in

wet ice

storage temp.

−70°C

Gene Information

human ... CIRBP(1153)

General description

CIRBP, also known as cold-inducible RNA-binding protein, plays a protective role in the genotoxic stress response by stabilizing transcripts of genes involved in cell survival. CIRBP responds to a wide array of cellular stresses, including short wavelength ultraviolet light (UVC), at the transcriptional and post-translational level. It acts as a translational activator. CIRBP can bind the 3′untranslated region of specific transcripts to stabilize them and facilitate their transport to ribosomes for translation. CIRBP affects NF-κB signaling as opposed to IL1B mRNA stability directly.

Physical form

Lyophilized from sterile 20 mM Tris, 500 mM NaCl, 0.25 M Imidazole, 20% glycerol.

Reconstitution

Detailed reconstitution instructions are sent along with the product

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service