Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP6529

Sigma-Aldrich

NAD Kinase human

recombinant, expressed in E. coli, ≥95% (SDS-PAGE)

Synonym(s):

EC 2.7.1.23, NADK, Poly (P)/ATP NAD Kinase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352204
NACRES:
NA.54

biological source

human

recombinant

expressed in E. coli

Assay

≥95% (SDS-PAGE)

form

liquid

mol wt

monomer ~50 kDa (monomer.Human full length NAD kinase aa 1- 446 is fused at the N-terminus to a His-tag.)

packaging

pkg of 10 μg

concentration

1 mg/mL

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... NADK(65220)

General description

NAD kinase is localized to mitochondria and cytosol. The gene is mapped to human chromosome 1p36.33.

Biochem/physiol Actions

NAD kinase EC 2.7.1.23 catalyzes the transfer of a phosphate group from ATP to NAD+ to generate NADP+, which acts as an electron donor for biosynthetic reactions upon reduction. NADP+ is an essential coenzyme in metabolism and provides reducing power to biosynthetic processes such as fatty acid biosynthesis.

Physical form

1 mg/mL of ammonium sulphate suspension.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Significance of Ser-188 in human mitochondrial NAD kinase as determined by phosphomimetic and phosphoresistant amino-acid substitutions.
Kawabata Y, et al.
Biochemical and Biophysical Research Communications, 468(4), 691-695 (2015)
Enzymology of NAD+ homeostasis in man.
Magni G, et al.
Cellular and Molecular Life Sciences, 61(1), 19-34 (2004)
Leonardo A Meza-Zepeda et al.
Journal of cellular and molecular medicine, 12(2), 553-563 (2008-04-19)
The potential use of human mesenchymal stem cells for therapeutic applications implies large scale in vitro culture, increasing the probability of genetic instability and transformation. We examine here the incidence of unbalanced and balanced chromosome rearrangements in polyclonal and single

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service