Skip to Content
MilliporeSigma
All Photos(1)

Documents

SRP6474

Sigma-Aldrich

GFRA1/GDNFRA human

recombinant, expressed in HEK 293 cells, ≥95% (SDS-PAGE)

Synonym(s):

GDNFR, GFR-ALPHA-1, RET1L, RETL1, TRNR1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in HEK 293 cells

tag

6-His tagged (C-terminus)

Assay

≥95% (SDS-PAGE)

form

lyophilized

mol wt

calculated mol wt 45.5 kDa
observed mol wt 55-60 kDa (DTT-reduced. Asp 25 is the predicted N-terminal.)

packaging

pkg of 10 and 50 μg

impurities

<1 EU/μg endotoxin (LAL test)

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... GFRA1(2674)

General description

GDNF family receptor α-1 (GFRA1) also known as RET ligand 1 (RETL1), TGF-β-related neurotrophic factor receptor 1 (TRNR1), is a novel glycosylphosphatidylinositol (GPI)-linked cell surface receptor, and belongs to the glial cell line-derived neurotrophic factor (GDNF) receptor subfamily. The GDNF family ligands comprises GDNF, Neurturin (NTN), Artemin, and Persephin. GDNF and NTN are two structurally related, potent neurotrophic factors that play key roles in the control of neuron survival and differentiation. GFRA1 mediates the association with and activation of the RET tyrosine kinase receptor (RTK), and subsequently initiates the RET signaling pathway. GFR α-mediated signaling plays an important role in the survival, differentiation, and migration of central and peripheral neurons, and is essential for the development of kidneys and the enteric nervous system, as well as the inflammatory response in some carcinomas.

Physical form

Lyophilized from 0.22 μm filtered solution in PBS, pH 7.4. Normally Mannitol or Trehalose is added as protectants before lyophilization.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in sterile PBS, pH 7.4 to a concentration of 50 μg/mL. Do not vortex. This solution can be stored at 2-8°C for up to 1 month. For extended storage, it is recommended to store at -20°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service