Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP6470

Sigma-Aldrich

CADM1 human

recombinant, expressed in HEK 293 cells, ≥95% (SDS-PAGE)

Synonym(s):

BL2, DKFZp686F1789, IGSF4, IGSF4A, MGC149785, NECL2, Necl-2, RA175, ST17, SYNCAM, TSLC1, sTSLC-1, sgIGSF, synCAM1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

biological source

human

recombinant

expressed in HEK 293 cells

Assay

≥95% (SDS-PAGE)

form

lyophilized

mol wt

calculated mol wt 38.5 kDa
observed mol wt 38.5_kDa,50_kDaand55-85 kDa
observed mol wt 50 kDa
observed mol wt 55-85 kDa

packaging

pkg of 10 μg
pkg of 50 μg

impurities

<1 EU/μg endotoxin (LAL test)

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... CADM1(23705)

General description

Cell Adhesion Molecules (CAMs) are proteins located on the cell surface involved with the binding with other cells or with the extracellular matrix (ECM) in the process called cell adhesion. Cell adhesion molecule 1 is a single-pass type I membrane protein which belongs to the nectin family. Cell Adhesion Molecule 1 contains two Ig-like C2-type (immunoglobulin-like) domains and one Ig-like V-type (immunoglobulin-like) domain, and is also known as CADM1; BL2; DKFZp686F1789; IGSF4; IGSF4A; MGC149785; MGC51880; NECL2; Necl-2; RA175; ST17; SYNCAM; TSLC1; sTSLC-1; sgIGSF; synCAM1. CADM1 mediates heterophilic cell-cell adhesion with CADM3 and PVRL3 in a Ca2+-independent manner. CADM1 acts as a tumor suppressor in non-small-cell lung cancer (NSCLC) cells. CADM1 may be involved in neuronal migration, axon growth, path finding, and fasciculation on the axons of differentiating neurons. CADM1 may play diverse roles in the spermatogenesis including in the adhesion of spermatocytes and spermatids to Sertoli cells and for their normal differentiation into mature spermatozoa.
DTT-reduced. SDS-PAGE, rhCADM1Protein migrates as 38.5 kDa, 50 kDa and 55-85 kDa poly peptide due to different glycosylation. Gln 45 is the predicted N-terminal.

Physical form

Lyophilized from 0.22 μm filtered solution in PBS, pH 7.4. Normally Mannitol or Trehalose is added as protectants before lyophilization.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in sterile PBS, pH 7.4 to a concentration of 50 μg/mL. Do not vortex. This solution can be stored at 2-8°C for up to 1 month. For extended storage, it is recommended to store at -20°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shingo Nakahata et al.
Haematologica, 106(2), 532-542 (2020-02-15)
Adult T-cell leukemia/leukemia (ATLL) is an aggressive peripheral T-cell malignancy, caused by infection with the human T-cell leukemia virus type 1 (HTLV-1). We have recently shown that cell adhesion molecule 1 (CADM1), a member of the immunoglobulin superfamily, is specifically

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service