Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP6469

Sigma-Aldrich

VLDLR human

recombinant, expressed in HEK 293 cells, ≥97% (SDS-PAGE)

Synonym(s):

CARMQ1, CHRMQ1, FLJ35024, VLDL-R, VLDLRCH, very-low-density-lipoprotein receptor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

biological source

human

recombinant

expressed in HEK 293 cells

tag

6-His tagged (C-terminus)

Assay

≥97% (SDS-PAGE)

form

lyophilized

mol wt

calculated mol wt 86 kDa
observed mol wt 150 kDa by SDS-PAGE (DTT-reduced.)
observed mol wt 180 kDa

packaging

pkg of 10 μg
pkg of 50 μg

impurities

<1 EU/μg endotoxin (LAL test)

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... VLDLR(7436)

General description

Very-low-density-lipoprotein receptor (VLDLR) is a 130 kDa type I transmembrane protein in the low density lipoprotein (LDL) receptor family that plays a significant role in lipid metabolism and in nervous system development and function.[1][2] The VLDL receptor structure is more or less homologous to that LDL receptor, excluding the eightfold repeated ligand binding domain of VLDLR, which is sevenfold in LDLR. The protein sequence of VLDLR is mostly conserved among mammals such as rabbit, human and mouse.[3] The gene is mapped to human chromosome 9p24.2.[4]

Biochem/physiol Actions

Very-low-density-lipoprotein receptor (VLDLR) is associated with the metabolism of triglyceride rich lipoproteins (VLDL,β-migrating VLDL and intermediate-density lipoprotein) involving apoprotein-E.[3] It is also one of the receptors of reelin, an extracellular matrix protein which regulates the processes of neuronal migration and synaptic plasticity. It is associated with parental consanguinity and found in secluded communities such as the Hutterites.[5][6] Deletion in the VLDLR gene results in cerebellar hypoplasia an autosomal recessive syndrome.[5] VLDLR gene is significantly associated with Reelin pathway.[2]

Physical form

Lyophilized from 0.22 μm filtered solution in PBS, pH 7.4. Normally Mannitol or Trehalose is added as protectants before lyophilization.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in sterile PBS, pH 7.4 to a concentration of 50 μg/mL. Do not vortex. This solution can be stored at 2-8°C for up to 1 month. For extended storage, it is recommended to store at -20°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mouse very?low?density?lipoprotein receptor (VLDLR) cDNA cloning, tissue?specific expression and evolutionary relationship with the low?density?lipoprotein receptor.
Oka K, et al.
European Journal of Biochemistry, 224(3), 975-982 (1994)
Reelin is a ligand for lipoprotein receptors.
D'Arcangelo G, et al.
Neuron, 24(2), 471-479 (1999)
Lipid Peroxidation Induced ApoE Receptor-Ligand Disruption as a Unifying Hypothesis Underlying Sporadic Alzheimer's Disease in Humans.
Ramsden, et al.
Journal of Alzheimer'S Disease, 87, 1251-1290 (2022)
Decreased expression of VLDLR is inversely correlated with miR?200c in human colorectal cancer.
Kim B K, et al.
Molecular Carcinogenesis, 56(6), 1620-1629 (2017)
Genomic loss and epigenetic silencing of very-low-density lipoprotein receptor involved in gastric carcinogenesis.
Takada H, et al.
Oncogene, 25(49), 6554-6554 (2006)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service