Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP6459

Sigma-Aldrich

EPOR/Erythropoietin Receptor human

recombinant, expressed in HEK 293 cells, ≥92% (SDS-PAGE)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

biological source

human

recombinant

expressed in HEK 293 cells

Assay

≥92% (SDS-PAGE)

form

lyophilized

mol wt

calculated mol wt 50.1 kDa (This protein is fused with Fc fragment of human IgG1 at the C-terminus.)
observed mol wt 50.1 kDa (DTT-reduced. Protein migrates due to glycosylation. Ala 25 is the predicted N-terminal.)

packaging

pkg of 10 and 50 μg

impurities

<1 EU/μg endotoxin (LAL test)

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... EPOR(2057)

General description

Erythropoietin (EPO) is the major glycoprotein hormone regulator of mammalian erythropoiesis, and is produced by kidney and liver in an oxygen-dependent manner. The biological effects of EPO are mediated by the specific erythropoietin receptor (EPOR) on bone marrow erythroblasts, which transmits signals important for both proliferation and differentiation along the erythroid lineage. EPOR is a type 1 single-transmembrane cytokine receptor, and belongs to the homodimerizing subclass which functions as ligand-induced or ligand-stabilized homodimers. EPOR pre-exists as dimers which upon binding of a 34 kDa ligand erythropoietin (EPO), changes its homodimerized state. These conformational changes result in the autophosphorylation of Jak2 kinases that are pre-associated with the receptor. Erythropoietin is necessary to maintain endothelial cells and to promote tumor angiogenesis, hence the dysregulation of EpoR may affect the growth of certain tumors. EpoR signaling prevents neuronal death and ischemic injury.

Physical form

Lyophilized from 0.22 μm filtered solution in 50 mM tris, 100 mM glycine, pH 7.0. Normally Mannitol or Trehalose is added as protectants before lyophilization.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in sterile PBS, pH 7.4 to a concentration of 50 μg/mL. Do not vortex. This solution can be stored at 2-8°C for up to 1 month. For extended storage, it is recommended to store at -20°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service