Skip to Content
MilliporeSigma
All Photos(1)

Documents

SRP6458

Sigma-Aldrich

TFPI human

recombinant, expressed in HEK 293 cells, ≥95% (SDS-PAGE)

Synonym(s):

EPI, LACI, TFI, TFPI1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

biological source

human

recombinant

expressed in HEK 293 cells

tag

6-His tagged (C-terminus)

Assay

≥95% (SDS-PAGE)

form

lyophilized

potency

<0.35 nM IC50

mol wt

calculated mol wt 30 kDa
observed mol wt 41-45 kDa (DTT-reduced. Protein migrates due to glycosylation. Asp 29 is the predicted N-terminal.)

packaging

pkg of 10 μg

impurities

<1 EU/μg endotoxin (LAL test)

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... TFPI(7035)

General description

The TFPI (tissue factor pathway inhibitor) gene is mapped to human chromosome 2q32.1. This protein exists in two isoforms TFPIα and TFPIβ.
Tissue factor pathway inhibitor (TFPI) also known as Extrinsic pathway inhibitor (EPI), Lipoprotein - associated coagulation inhibitor (LACI), is a plasma proteinase inhibitor synthesized by vascular endothelial cells and part of it is associated with glycosaminoglycans of these cells. TFPI is a single-chain polypeptide which can reversibly inhibit Factor Xa (Xa) and Thrombin (Factor IIa). TFPI is a secreted protein with a N­terminal acidic region, three Kunitz (K) domains separated with by two linker regions, and a C­terminal basic region. The first K domain inhibits coagulation factor VIIa complexed to tissue factor (TF); The second K domain inhibits factor Xa; The third K domain binds to heparin; The C­terminal basic region may have several functions. For example, it plays an important role in binding of TFPI to cell surfaces.

Biochem/physiol Actions

Inhibition of tissue factor function and thrombus formation by TFPI (tissue factor pathway inhibitor) is a major reason for ischemic stroke in carotid artery disease. The main function of TFPI is to regulate blood coagulation system. It possess both anticoagulant and antimetastatic action. This protein is prominently localized to medial and neointimal smooth muscle cells, macrophages and T-cells of atherosclerotic plaques. Upregulation of the TFPI gene is observed in atherosclerosis. Mutation in the TFPI gene leads to the risk of coronary artery disease.

Physical form

Lyophilized from 0.22 μm filtered solution in PBS, pH 7.4. Normally Mannitol or Trehalose is added as protectants before lyophilization.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in sterile PBS, pH 7.4 to a concentration of 50 μg/mL. Do not vortex. This solution can be stored at 2-8°C for up to 1 month. For extended storage, it is recommended to store at -20°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Teresa C Assumpção et al.
PLoS neglected tropical diseases, 10(1), e0004298-e0004298 (2016-01-09)
Hematophagous mosquitos and ticks avoid host hemostatic system through expression of enzyme inhibitors targeting proteolytic reactions of the coagulation and complement cascades. While most inhibitors characterized to date were found in the salivary glands, relatively few others have been identified
Significant genetic association of a functional TFPI variant with circulating fibrinogen levels and coronary artery disease
Naji DH, et al.
Molecular Genetics and Genomics, 293(1), 119-128 (2018)
Increased expression of TFPI in human carotid stenosis
Stavik B, et al.
Thrombosis Research, 155(5), 31-37 (2017)
John W Avery et al.
PloS one, 7(2), e31090-e31090 (2012-02-22)
Low birth weight and fetal loss are commonly attributed to malaria in endemic areas, but the cellular and molecular mechanisms that underlie these poor birth outcomes are incompletely understood. Increasing evidence suggests that dysregulated hemostasis is important in malaria pathogenesis
Tissue Factor Pathway Inhibitor Gene Polymorphism- 33T C Predicts Improved Disease-Free Survival in Colorectal Cancer
Bazzarell AK, et al.
Annals of Surgical Oncology, 23(7), 2274-2280 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service