Skip to Content
MilliporeSigma
All Photos(1)

Documents

SRP6439

Sigma-Aldrich

CD62E/E-Selectin human

recombinant, expressed in HEK 293 cells, ≥95% (SDS-PAGE)

Synonym(s):

E-Selectin, ELAM1, ESEL, LECAM2, RP1-117P20.2, SELE

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

biological source

human

recombinant

expressed in HEK 293 cells

Assay

≥95% (SDS-PAGE)

form

lyophilized

mol wt

calculated mol wt 60 kDa
observed mol wt 110 kDa by SDS-PAGE (DTT-reduced. Trp22 is the predicted N-terminal.)

packaging

pkg of 100 μg
pkg of 20 μg

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... SELE(6401)

General description

Selectins are a family of calcium-dependent type 1 transmembrane proteins. Endothelial (E)-selectin is a heavily glycosylated transmembrane protein expressed by activated endothelial cells in microvascular linings. E-selectin consists of an N-terminal type 1 lectin domain, an EGF-like domain, 6 sushi (CCP/SCR) domains, a transmembrane sequence, and a short cytoplasmic domain. Recombinant human E-selectin is a 58.6kDa protein containing 535 amino acid residues, corresponding to the extracellular portion of the full length protein. Due to glycosylation, E-selectin migrates at an apparent molecular weight of approximately 65-85kDa by SDS-PAGE analysis under reducing conditions. The gene encoding this protein is localized on human chromosome 1q24.2.

Biochem/physiol Actions

E-selectin, along with P-selectin and L-selectin, initiate recruitment of circulating leukocytes from blood to sites of inflammation in the vascular lining through interaction with specific cell surface associated carbohydrate determinants. It is a pro-angiogenic factor. This protein has also been implicated in the pathogenesis of stroke and atherosclerosis.

Physical form

Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally Mannitol or Trehalose is added as protectants before lyophilization.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in sterile PBS, pH 7.4 to a concentration of 50 μg/mL. Do not vortex. This solution can be stored at 2-8°C for up to 1 month. For extended storage, it is recommended to store at -20°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E-selectin gene polymorphisms and essential hypertension in Asian population: an updated meta-analysis.
Cai G
PLoS ONE, 9(7), e102058-e102058 (2014)
Remodeling of the lectin-EGF-like domain interface in P- and L-selectin increases adhesiveness and shear resistance under hydrodynamic force.
Phan UT
Nature Immunology, 7(8), 883-889 (2006)
Two new susceptibility loci 1q24.2 and 11p11.2 confer risk to severe acne.
He L
Nature Communications, 5, 2870-2870 (2014)
Association of E-selectin gene polymorphism (S128R) with ischemic stroke and stroke subtypes.
Roy S
Inflammation, 37(2), 599-603 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service