Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP6388

Sigma-Aldrich

LAMP1/CD107a human

recombinant, expressed in HEK 293 cells, ≥95% (SDS-PAGE)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in HEK 293 cells

tag

6-His tagged (C-terminus)

Assay

≥95% (SDS-PAGE)

form

lyophilized

mol wt

calculated mol wt 39.2 kDa
observed mol wt 60-100 kDa (DTT-reduced. Protein migrates due to different glycosylation. Ala 29 is the predicted N-terminus.)

packaging

pkg of 10 and 50 μg

impurities

<1 EU/μg endotoxin (LAL test)

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... LAMP1(3916)

General description

Lysosomal-associated membrane protein 1 (LAMP1) is also known as CD107a (Cluster of Differentiation 107a). LAMP1 / CD107a belongs to the LAMP family. This glycoprotein provides selectins with carbohydrate ligands. It may also play a role in tumor cell metastasis. CD107a has also been shown to be a marker of degranulation on lymphocytes such as CD8+ and NK cells.

Physical form

Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Generally 5-8% Mannitol or trehalose is added as a protectant before lyophilization.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in sterile PBS, pH 7.4 to a concentration of 50 μg/mL. Do not vortex. This solution can be stored at 2-8°C for up to 1 month. For extended storage, it is recommended to store at -20°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service