Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP6346

Sigma-Aldrich

TREM1 human

recombinant, expressed in E. coli, ≥85% (SDS-PAGE)

Synonym(s):

CD354, TREM-1, Triggering receptor expressed on myeloid cells 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

Assay

≥85% (SDS-PAGE)

form

liquid

mol wt

23.3 kDa (209 aa, 21-205 aa + His Tag)

packaging

pkg of 100 μg

concentration

0.5 mg/mL

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... TREM1(54210)

General description

TREM1 is a receptor belonging to the Ig super family that is expressed on myeloid cells. This protein amplifies neutrophil and monocyte-mediated inflammatory responses triggered by bacterial and fungal infections by stimulating release of pro-inflammatory chemokines and cytokines, as well as increased surface expression of cell activation markers. Alternatively spliced transcript variants encoding different isoforms have been noted for this gene. Recombinant human TREM1 protein, fused to His-tag at N-terminus, was expressed in E.coli and purified by using conventional chromatography techniques.

Physical form

0.5 mg/mL in 20 mM Tris-HCl buffer (pH 8.0) containing 0.15 M NaCl, 30% glycerol and 1 mM DTT.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service