Skip to Content
MilliporeSigma
All Photos(1)

Documents

SRP6300

Sigma-Aldrich

Apolipoprotein A-II from human plasma

≥95% (SDS-PAGE)

Synonym(s):

APOA2, Apo-AII, ApoA-II, Apolipoprotein A-II

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

biological source

human plasma

Assay

≥95% (SDS-PAGE)

form

frozen liquid

mol wt

17.4 kDa

packaging

pkg of 500 μg

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... ApoA2(336)

General description

APOA2 stabilizes the HDL structure by its relationship with lipids, and plays a role in the HDL metabolism. APOA2 is the 2nd most abundant protein of the high density lipoprotein particles. 25% of Apo A in HDL is Apo AII. APOA2 is localized in plasma as a monomer, homodimer, or heterodimer with apolipoprotein D. Defects in APOA2 gene might cause apolipoprotein A-II deficiency or hypercholesterolemia. Levels of Apo AII in normal plasma range from 30-50 mg/100 mL. The physiological role of the protein is unknown. Alcohol consumption, however, increases Apo AII levels.

Physical form

Frozen in 10 mM NH4HCO3, pH 7.4.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service