Skip to Content
MilliporeSigma
All Photos(1)

Documents

SRP6264

Sigma-Aldrich

ADAM12 human

recombinant, expressed in HEK 293 cells, ≥95% (SDS-PAGE)

Synonym(s):

ADAM12, MCMP, MCMPMltna, MLTN, MLTNA, Meltrin Alpha

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in HEK 293 cells

tag

6-His tagged (C-terminus)

Assay

≥95% (SDS-PAGE)

form

lyophilized powder

mol wt

calculated mol wt 34 kDa (Mature)
calculated mol wt 55.2 kDa
observed mol wt 23 kDa
observed mol wt 46 kDa
observed mol wt 65 kDa

packaging

pkg of 10 μg

impurities

<1 EU/μg endotoxin (LAL test)

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... ADAM12(8038)

General description

A Disintegrin And Metalloprotease 12 (ADAM12) is also known as Meltrin-α and MCMP, is a member of the ADAM and MDC (Metalloprotease, Disintegrin, Cysteine-rich) protein family. Members of this family are membrane-anchored proteins structurally related to snake venom disintegrins, and have been implicated in a variety of biological processes involving in diverse processes such as asthma, development, angiogenesis and cancer through their activities in cell adhesion/fusion, membrane protein shedding, and signal transduction. Over 30 members have been identified and about half of them are active metalloproteases such as ADAM8, 9, 10, 12 and 17/TACE. ADAM12 has four Isoform, Isoform 1 is expressed in placenta and skeletal, cardiac, and smooth muscle. Isoform 2 seems to be expressed only in placenta or in embryo and fetus. Both forms were expressed in some tumor cells lines. ADAM12 Involved in skeletal muscle regeneration, specifically at the onset of cell fusion and also involved in macrophage-derived giant cells (MGC) and osteoclast formation from mononuclear precursors.
DTT-reduced. Protein migrates due to glycosylation corresponding to the propeptide, the mature form and the pro form respectively. Leu24 is the predicted N-terminus.

Physical form

Lyophilized from 0.22 μm filtered solution in PBS. Generally 5-8% Mannitol or trehalose is added as a protectant before lyophilization.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in sterile PBS, pH 7.4 to a concentration of 50 μg/mL. Do not vortex. This solution can be stored at 2-8°C for up to 1 month. For extended storage, it is recommended to store at -20°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Uma S Kamaraj et al.
Cell systems, 11(5), 509-522 (2020-10-11)
The need to derive and culture diverse cell or tissue types in vitro has prompted investigations on how changes in culture conditions affect cell states. However, the identification of the optimal conditions (e.g., signaling molecules and growth factors) required to maintain

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service