Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP6260

Sigma-Aldrich

SERPINA1/A1AT human

recombinant, expressed in HEK 293 cells, ≥95% (SDS-PAGE)

Synonym(s):

A1A, A1AT, AAT, MGC23330, MGC9222, PI, PI1, PRO2275, SPAAT, SerpinA1, alpha1AT

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in HEK 293 cells

tag

6-His tagged (C-terminus)

Assay

≥95% (SDS-PAGE)

form

lyophilized powder

potency

<4.5 nM IC50

mol wt

calculated mol wt 45.1 kDa
observed mol wt 55-60 kDa (DTT-reduced. Protein migrates due to glycosylation. Glu 25 is the predicted N-terminus.)

packaging

pkg of 10 μg

impurities

<1 EU/μg endotoxin (LAL test)

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... SERPINA1(5265)

General description

Serpin A1 also known as α-1-antitrypsin (A1AT), serum trypsin inhibitor, α-1 proteinase inhibitor (A1PI), AAT, which belongs to the serpin family. Most serpins inactivate enzymes by binding to them covalently, requiring very high levels to perform their function. Like all serine protease inhibitors, A1AT has a characteristic secondary structure of β sheets and α helices. Serpin A1 / A1AT is inhibitor of serine proteases. Its primary target is elastase, but it also has a moderate affinity for plasmin and thrombin. Irreversibly inhibits trypsin, chymotrypsin and plasminogen activator. The aberrant form of SerpinA1 inhibits insulin-induced NO synthesis in platelets, decreases coagulation time and has proteolytic activity against insulin and plasmin. Serpin A1 / A1AT protects tissues from enzymes of inflammatory cells, especially neutrophil elastase. Defects in SERPINA1 are the cause of α-1-antitrypsin deficiency (A1ATD).

Physical form

Lyophilized from 0.22 μm filtered solution in PBS. Generally 5-8% Mannitol or trehalose is added as a protectant before lyophilization.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in sterile PBS, pH 7.4 to a concentration of 50 μg/mL. Do not vortex. This solution can be stored at 2-8°C for up to 1 month. For extended storage, it is recommended to store at -20°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service