Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SRP6256

Sigma-Aldrich

VEGF-C human

recombinant, expressed in HEK 293 cells, ≥97% (SDS-PAGE)

Synonym(s):

Flt4-L, VRP

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

biological source

human

recombinant

expressed in HEK 293 cells

tag

6-His tagged (C-terminus)

Assay

≥97% (SDS-PAGE)

form

lyophilized powder

potency

0.15-0.7 μg/mL

mol wt

calculated mol wt 15 kDa
observed mol wt 18-23 kDa (DTT-reduced. Protein migrates due to glycosylation. Thr 103 is the predicted N-terminus .)

packaging

pkg of 10 μg

impurities

<1 EU/μg endotoxin (LAL test)

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... VEGF-C(7424)

General description

Vascular endothelial growth factor C is also known as VEGFC, Flt4-L and VRP; it contains the C-terminal propeptide which has an unusual structure with tandemly repeated cysteine-rich motifs. Upon biosynthesis, VEGFC is secreted as a non-covalent momodimer in an anti-parallel fashion. VEGFC is a member of the platelet-derived growth factor/vascular endothelial growth factor (PDGF/VEGF) family, is active in angiogenesis, lymphangiogenesis and endothelial cell growth and survival, and can also affect the permeability of blood vessels. This secreted protein undergoes a complex proteolytic maturation, generating multiple processed forms that bind and activate VEGFR-3 receptors. Only the fully processed form can bind and activate VEGFR-2 receptors. The structure and function of this protein is similar to those of vascular endothelial growth factor D (VEGF-D). VEGFC may function in angiogenesis of the venous and lymphatic vascular systems during embryogenesis, and also in the maintenance of differentiated lymphatic endothelium in adults. Overexpression of VEGF-C causes lymphatics to enlarge possibly facilitates metastasis.

Physical form

Lyophilized from 0.22 μm filtered solution in PBS. Generally 5-8% Mannitol or trehalose is added as a protectant before lyophilization.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in sterile PBS, pH 7.4 to a concentration of 50 μg/mL. Do not vortex. This solution can be stored at 2-8°C for up to 1 month. For extended storage, it is recommended to store at -20°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service