Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP6226

Sigma-Aldrich

Nesfatin-1 human

recombinant, expressed in E. coli, ≥98% (SDS-PAGE)

Synonym(s):

Nesfatin 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

Assay

≥98% (SDS-PAGE)

form

lyophilized powder

mol wt

9.7 kDa

packaging

pkg of 10 and 50 μg

impurities

<1 EU/μg endotoxin (LAL test)

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... NEFA(4925)

General description

Nesfatin-1 is a metabolic polypeptide encoded in the N-terminal region of the precursor protein, Nucleobindin2 (NUCB2). Originally identified as a hypothalamic neuropeptide, Nesfatin-1 is also expressed in other areas of the brain, and in pancreatic islets β-cells, gastric endocrine cells and adipocytes. Nesfatin-1 suppresses food intake and can regulate energy metabolism in a Leptin independent manner. Recombinant human Nesfatin-1 is a 9.7 kDa protein containing 82 amino acid residues.

Physical form

Sterile filtered through a 0.2 micron filter. Lyophilized from 0.1% TFA.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/mL. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service