Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP6201

Sigma-Aldrich

Amphiregulin human

recombinant, expressed in E. coli, ≥95% (SDS-PAGE)

Synonym(s):

AR, AREG, Colorectum cell-derived growth factor (CRDGF)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

Assay

≥95% (SDS-PAGE)

form

lyophilized powder

mol wt

11.3 kDa

packaging

pkg of 10 μg
pkg of 50 μg

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... AREG(374)

General description

Amphiregulin is an EGF related growth factor that signals through the EGF/TGF-a receptor, and stimulates growth of keratinocytes, epithelial cells and some fibroblasts. Amphiregulin also inhibits the growth of certain carcinoma cell lines. Synthesized as a transmembrane protein, amphiregulin′s extracellular domain is proteolytically processed to release the mature protein. There are 6 conserved cysteine residues, which form 3 intramolecular disulfide bonds essential for biological activity. Recombinant human Amphiregulin is a 11.3 kDa glycoprotein consisting of 98 amino acid residues.

Physical form

Sterile filtered through a 0.2 micron filter. Lyophilized from 5 mM Sodium phosphate, pH 7.0.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/mL. Do not vortex. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michaela Pommer et al.
Cells, 10(2) (2021-02-11)
Oncogene-induced senescence (OIS) is a decisive process to suppress tumor development, but the molecular details of OIS are still under investigation. Using an established OIS model of primary melanocytes transduced with BRAF V600E and compared to control cells, amphiregulin (AREG)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service