Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP6186

Sigma-Aldrich

Platelet Factor 4 from human platelets

≥95% (SDS-PAGE)

Synonym(s):

C-X-C motif chemokine 4, Iroplact, Oncostatin-A

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202

biological source

human

Assay

≥95% (SDS-PAGE)

form

lyophilized

mol wt

27.1 kDa

packaging

pkg of 20 μg

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... PF4(5196)

General description

A heparin-neutralizing protein which is contained in platelet storage granules. Platelet Factor 4 is released during aggregation, but its actual role in normal or abnormal hemostasis is unknown.

Physical form

Lyophilized from 20 mM Tris, pH 8.3, with 1.5 M NaCl.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service