Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP6185

Sigma-Aldrich

Factor Va from human plasma

≥95% (SDS-PAGE)

Synonym(s):

Activated protein C cofactor, Proaccelerin (labile factor)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

biological source

human plasma

Assay

≥95% (SDS-PAGE)

form

liquid

mol wt

heavy chain 94.0 kDa
light chain 74 kDa

packaging

pkg of 10 μg

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... F5(2153)

General description

Factor Va is a cofactor for the serine protease factor Xa, and in the presence of calcium ions they collectively assemble on a phospholipid surface to form the prothrombinase complex. The prothrombinase complex is responsible for the rapid conversion of prothrombin to thrombin. Factor Va is derived from the pro-cofactor, factor V, upon limited proteolysis by α-thrombin. The thrombin cleavage of factor V liberates two heavily glycosylated activation peptides from the central portion of the molecule which have no cofactor function. Factor Va is comprised of an NH2-terminal derived heavy chain (Mr=94,000) and a COOH-terminal derived light chain (Mr=74,000) which remain associated in the presence of calcium ions. The cofactor binds to phospholipid (cell membrane) surfaces and effectively serves as a receptor for membrane bound factor Xa. Complete assembly of the prothrombinase complex (factor Xa, factor Va, phospholipid, and calcium) results in a 300,000-fold increase in the rate of prothrombin conversion relative to the rate observed with factor Xa alone. The interaction between factor Va and factor Xa is mediated by both the heavy and light chain of factor Va, while the binding of prothrombin to factor Va is mediated solely by the heavy chain.

Physical form

liquid in 41.3 μl 20 mM HEPES, 150 mM NaC1, pH 7.5, 5 mM CaCl2 +50% glycerol.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service