Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP6124

Sigma-Aldrich

SORD human

recombinant, expressed in E. coli, ≥90% (SDS-PAGE)

Synonym(s):

L-iditol 2-dehydrogenase, Sorbitol dehydrogenase

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

CAS Number:
UNSPSC Code:
12352202

biological source

human

recombinant

expressed in E. coli

Assay

≥90% (SDS-PAGE)

form

liquid

mol wt

40.4 kDa (377 aa, 1-357 aa + NT His Tag)

packaging

pkg of 100 μg

storage condition

avoid repeated freeze/thaw cycles

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... SORD(6652)

General description

SORD, also known as L-iditol 2-dehydrogenase or SORD1, is a 357 amino acid member of the zinc containing alcohol dehydrogenase family. It is widely expressed with highest expression in kidney and in the lens of the eye. SORD enzymatically catalyzes the zinc-dependent interconversion of polyols, such as sorbitol and xylitol, to their respective ketoses.

Physical form

0.5 mg/mL solution in 20 mM Tris-HCl buffer (pH 8.0) containing 0.2 M NaCl, 5 mM DTT and 20% glycerol.

Preparation Note

Centrifuge the vial prior to opening.

Other Notes

MGSSHHHHHH SSGLVPRGSH MAAAAKPNNL SLVVHGPGDL RLENYPIPEP GPNEVLLRMH SVGICGSDVH YWEYGRIGNF IVKKPMVLGH EASGTVEKVG SSVKHLKPGD RVAIEPGAPR ENDEFCKMGR YNLSPSIFFC ATPPDDGNLC RFYKHNAAFC YKLPDNVTFE EGALIEPLSV GIHACRRGGV TLGHKVLVCG AGPIGMVTLL VAKAMGAAQV VVTDLSATRL SKAKEIGADL VLQISKESPQ EIARKVEGQL GCKPEVTIEC TGAEASIQAG IYATRSGGTL VLVGLGSEMT TVPLLHAAIR EVDIKGVFRY CNTWPVAISM LASKSVNVKP LVTHRFPLEK ALEAFETFKK GLGLKIMLKC DPSDQNP

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Miguel A Lanaspa et al.
Nature communications, 4, 2434-2434 (2013-09-12)
Carbohydrates with high glycaemic index are proposed to promote the development of obesity, insulin resistance and fatty liver, but the mechanism by which this occurs remains unknown. High serum glucose concentrations are known to induce the polyol pathway and increase

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service