Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP6121

Sigma-Aldrich

AK1 human

recombinant, expressed in E. coli, ≥95% (SDS-PAGE)

Synonym(s):

no change, ATP-AMP phophotransferase, ATP-AMP transphosphorylase 1, Adenylate kinase isoenzyme 1, Adenylate monophosphate kinase, Myokinase

Sign Into View Organizational & Contract Pricing


About This Item

Enzyme Commission number:
UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

Assay

≥95% (SDS-PAGE)

form

liquid

mol wt

23.7 kDa (214 aa, 1-194 aa + NT His Tag)

purified by

chromatography

packaging

pkg of 100 μg

technique(s)

activity assay: suitable

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... AK1(203)

General description

Research area: Cancer


The gene AK1 (adenylate kinase isoenzyme 1) is mapped to human chromosome 9 on the location 9q34.11.[1] It belongs to the nucleotide monophosphate kinase family.[2] The AK1 protein is found in the cytosol of skeletal muscle, brain, and heart.[3]The structure of AKs (adenylate kinases) comprises of three functional domains: the catalytic and oligomerization regulatory enzyme (CORE) domain, the ligand interaction domain (LID) domain, and the substrate binding site. In human tissues, a total of nine distinct isoenzymes of adenylate kinase are known, namely AK1 to AK9. [4] Recombinant human AK1 protein, fused to His-tag at N-terminus, was expressed in Escherichia coli and purified by using conventional chromatography.

Biochem/physiol Actions

AK1 (adenylate kinase isoenzyme 1) is an enzyme involved in regulating the adenine nucleotide composition within a cell by catalyzing the reversible transfer of the terminal phosphate group between ATP and AMP.[3] Low levels of AK1 cause imbalance in adenine nucleotide pools.[5] Defects in AK1 are the cause of a form of hemolytic anemia.[6] [1] This gene is also upregulated in the brains of AD (Alzheimer′s disease) patients and downregulated in Duchenne muscular dystrophy.[1] It is responsible for abnormal phosphorylation of tau via AMPK-GSK3β (AMP-activated protein kinase-glycogen synthase kinase-3β), thereby causing tau-mediated neurodegeneration.[7]AK1 plays an important role in the regulation of cancer cell metabolism, metabolic signaling, as well as cell migration and invasion. It also aids in the progression of malignant transformation.[8]

Physical form

1 mg/mL solution in 20 mM Tris-HCl buffer (pH 7.5) containing 10% glycerol.

Preparation Note

Centrifuge the vial prior to opening.

Other Notes

MGSSHHHHHH SSGLVPRGSH MEEKLKKTKI IFVVGGPGSG KGTQCEKIVQ KYGYTHLSTG DLLRSEVSSG SARGKKLSEI MEKGQLVPLE TVLDMLRDAM VAKVNTSKGF LIDGYPREVQ QGEEFERRIG QPTLLLYVDA GPETMTQRLL KRGETSGRVD DNEETIKKRL ETYYKATEPV IAFYEKRGIV RKVNAEGSVD SVFSQVCTHL DALK

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Defective metabolic signaling in adenylate kinase AK1 gene knock-out hearts compromises post-ischemic coronary reflow.
Dzeja PP, et al.
The Journal of Biological Chemistry, 282, 31366-31366 (2007)
Neuropathogenic role of adenylate kinase-1 in A?-mediated tau phosphorylation via AMPK and GSK3?.
Park H, et al.
Human Molecular Genetics, 21, 2725-2725 (2012)
The many isoforms of human adenylate kinases
Panayiotou C, et al.
The International Journal of Biochemistry & Cell Biology, 49, 75-83 (2014)
Metabolism of circulating ADP in the bloodstream is mediated via integrated actions of soluble adenylate kinase-1 and NTPDase1/CD39 activities.
Yegutkin GG, et al.
Faseb Journal, 26, 3875-3875 (2012)
Adenylate Kinase: A Ubiquitous Enzyme Correlated with Medical Conditions
Mihaela Ileana Ionescu
The Protein Journal, 38(2), 120-133 (2019)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service