Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP6105

Sigma-Aldrich

MDH from bacteria

recombinant, expressed in E. coli, ≥95% (SDS-PAGE)

Synonym(s):

ECK3225, JW3205, Malate dehydrogenase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352204
NACRES:
NA.32

biological source

bacterial

recombinant

expressed in E. coli

Assay

≥95% (SDS-PAGE)

form

liquid

mol wt

34.9 kDa (336 aa, 1-312 aa + NT His Tag)

packaging

pkg of 100 μg

technique(s)

activity assay: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

Escherichia coli ... MDH(947854)

General description

Malate dehydrogenase (MDH) belongs to the LDH/MDH superfamily and MDH type 1 family. It exists as a homodimer and tetramer in prokaryotes.[1]

Biochem/physiol Actions

Malate dehydrogenase (MDH) catalyzes the conversion of malate into oxaloacetate (using NAD+) and vice versa (reversible reaction). This reaction is part of many metabolic pathways, including the citric acid cycle. MDH has a key role in the central oxidative pathway. Malate dehydrogenase is also involved in gluconeogenesis, the synthesis of glucose from smaller molecules.[1]

Physical form

1 mg/mL solution in 20 mM Tris-HCl (pH 8.0) containing 1 mM DTT, 50 mM NaCl and 10% glycerol.

Preparation Note

Centrifuge the vial prior to opening.

Other Notes

MGSSHHHHHH SSGLVPRGSH MGSHMKVAVL GAAGGIGQAL ALLLKTQLPS GSELSLYDIA PVTPGVAVDL SHIPTAVKIK GFSGEDATPA LEGADVVLIS AGVARKPGMD RSDLFNVNAG IVKNLVQQVA KTCPKACIGI ITNPVNTTVA IAAEVLKKAG VYDKNKLFGV TTLDIIRSNT FVAELKGKQP GEVEVPVIGG HSGVTILPLL SQVPGVSFTE QEVADLTKRI QNAGTEVVEA KAGGGSATLS MGQAAARFGL SLVRALQGEQ GVVECAYVEG DGQYARFFSQ PLLLGKNGVE ERKSIGTLSA FEQNALEGML DTLKKDIALG EEFVNK

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Priscila Pini Zenatti et al.
EBioMedicine, 30, 158-166 (2018-03-20)
The drug l-asparaginase is a cornerstone in the treatment of acute lymphoblastic leukemia (ALL). The native E. colil-asparaginase used in Brazil until recently has been manufactured by Medac/Kyowa. Then a decision was taken by the Ministry of Health in 2017
Function, kinetic properties, crystallization, and regulation of microbial malate dehydrogenase*
Toshiko T
Journal of Zhejiang University. Science. B, 17(4), 247-261 (2016)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service