Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP6081

Sigma-Aldrich

TXNRD1 human

recombinant, expressed in E. coli, ≥95% (SDS-PAGE)

Synonym(s):

GRIM-12, MGC9145, TR1, TRXR1, Thioredoxin reductase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

Assay

≥95% (SDS-PAGE)

form

liquid

potency

0.48 units/mL

mol wt

55.7 kDa

packaging

pkg of 100 μg

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... TXNRD1(7296)

General description

Thioredoxin reductase 1 (TNXRD1), cytoplasmic is an enzyme in humans that is encoded by the TXNRD1 gene. This gene encodes a member of the family of pyridine nucleotide oxidoreductases and is localized on human chromosome 12.

Biochem/physiol Actions

Thioredoxin reductase 1 (TNXRD1) reduces thioredoxins as well as other substrates. It plays a role in selenium metabolism and protection against oxidative stress. The functional enzyme is thought to be a homodimer which uses flavin adenine dinucleotide (FAD) as a cofactor. Inhibition of TXNRD1 activity may provide for potential treatments of cancer, acquired immune deficiency syndrome (AIDS), autoimmune diseases as well as bacterial infections and parasitic diseases.

Physical form

0.5 mg/mL solution in PBS (pH 7.4) containing 20% glycerol.

Preparation Note

Centrifuge the vial prior to opening.

Other Notes

MGSSHHHHHH SSGLVPRGSH MYDYDLIIIG GGSGGLAAAK EAAQYGKKVM VLDFVTPTPL GTRWGLGGTC VNVGCIPKKL MHQAALLGQA LQDSRNYGWK VEETVKHDWD RMIEAVQNHI GSLNWGYRVA LREKKVVYEN AYGQFIGPHR IKATNNKGKE KIYSAERFLI ATGERPRYLG IPGDKEYCIS SDDLFSLPYC PGKTLVVGAS YVALECAGFL AGIGLDVTVM VRSILLRGFD QDMANKIGEH MEEHGIKFIR QFVPIKVEQI EAGTPGRLRV VAQSTNSEEI IEGEYNTVML AIGRDACTRK IGLETVGVKI NEKTGKIPVT DEEQTNVPYI YAIGDILEDK VELTPVAIQA GRLLAQRLYA GSTVKCDYEN VPTTVFTPLE YGACGLSEEK AVEKFGEENI EVYHSYFWPL EWTIPSRDNN KCYAKIICNT KDNERVVGFH VLGPNAGEVT QGFAAALKCG LTKKQLDSTI GIHPVCAEVF TTLSVTKRSG ASILQAGC

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Birgitte Zeuner et al.
Enzyme and microbial technology, 82, 42-50 (2015-12-18)
Sialylated galactooligosaccharides (GOS) represent a potential infant formula ingredient, which is believed to contribute with a combination of the beneficial properties of the prebiotic GOS as well as of sialylated human milk oligosaccharides. Sialylated GOS do not exist in natural
Inés Mármol et al.
Pharmaceutics, 14(10) (2022-10-28)
In this paper we describe the synthesis of new N-heterocyclic carbene (NHC) gold(I) derivatives with flavone-derived ligands with a propargyl ether group. The compounds were screened for their antimicrobial and anticancer activities, showing greater activity against bacteria than against colon
Javier Quero et al.
Biomedicines, 9(12) (2021-12-25)
Overheating can affect solubility or lipophilicity, among other properties, of some anticancer drugs. These temperature-dependent changes can improve efficiency and selectivity of the drugs, since they may affect their bioavailability, diffusion through cell membrane or activity. One recent approach to
Nicole Swope et al.
Biotechnology and bioengineering, 117(4), 1063-1071 (2020-01-14)
Antibody disulfide bond (DSB) reduction during manufacturing processes is a widely observed phenomenon attributed to host cell reductases present in harvest cell culture fluid. Enzyme-induced antibody reduction leads to product fragments and aggregates that increase the impurity burden on the
Fine mapping and identification of a candidate gene SSH1 in disseminated superficial actinic porokeratosis.
Zhang Z
Human Mutation, 24(5), 438-438 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service