Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP6044

Sigma-Aldrich

Angiostatin from human plasma

Synonym(s):

Angiostatin, Angiostatin K1-4, Angiostatin Kringles 1-4

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

Assay

≥95%

form

liquid

mol wt

50.0 kDa

packaging

pkg of 100 μg

impurities

Endotoxin, tested

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... PLG(5340)

Physical form

In 20 mM HEPES, 150 mM NaCl, pH 7.4

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alvaro Macieira-Coelho
Inhibitors of Cell Growth (2012)
Angiostatin: an endogenous inhibitor of angiogenesis and of tumor growth.
O' Reilly MS
EXS (1997)
Characterization of a reduced form of plasma plasminogen as the precursor for angiostatin formation.
Butera D
The Journal of Biological Chemistry (2014)
Anti-angiogenesis mediated by angiostatin K1-3, K1-4 and K1-4.5. Involvement of p53, FasL, AKT and mRNA deregulation.
Chen YH
Thrombosis and Haemostasis (2006)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service