Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP6027

Sigma-Aldrich

UbcH5b human

recombinant, expressed in E. coli, ≥95% (SDS-PAGE)

Synonym(s):

UbcH

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

Assay

≥95% (SDS-PAGE)

form

liquid

mol wt

17.0 kDa

packaging

pkg of 10 μg

concentration

~1.0 mg/mL

impurities

Endotoxin, tested

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... UB2D2(7322)

General description

E2 ubiquitin conjugating enzyme (UbcH5b), also called as ubiquitin conjugating enzyme E2D2 (UBE2D2), belongs to the active class of E2 ubiquitin conjugating enzymes, which also includes UbcH5A and UbcH5C. UbcH5b structure contains four N-terminal α-helices followed by four-stranded antiparallel β-sheet.

Biochem/physiol Actions

E2 ubiquitin conjugating enzyme (UbcH5b) is a vital component of E3 complex SCFβ-TrCP and it plays an important role in phospho-IκBα ubiquitination. UbcH5b along with others members of active classes of E2 enzymes plays a vital role in degradation of many regulatory and abnormal proteins, including important human transcription factors, such as p53, nuclear factor-κB (NF-κB) and c-Fos.

Physical form

Supplied at a concentration of 1.0 mg/ml in 20 mM Tris-HCl, pH 7.5, containing 0.5 mM DTT.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Solution structure of the ubiquitin-conjugating enzyme UbcH5B.
Houben K
Journal of Molecular Biology, 344(2), 513-526 (2004)
The Shigella flexneri effector OspG interferes with innate immune responses by targeting ubiquitin-conjugating enzymes.
Kim DW
Proceedings of the National Academy of Sciences of the USA, 102(39), 14046-14051 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service