Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP5341

Sigma-Aldrich

ATG13, GST tagged human

recombinant, expressed in E. coli, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

KIAA0652

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
$472.00

$472.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
20 μG
$472.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

$472.00


Please contact Customer Service for Availability

Request a Bulk Order

recombinant

expressed in E. coli

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

mol wt

~96 kDa

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... ATG13(9776)

General description

ATG13 is a protein that is part of the autophagy complex which is the major route by which cytoplasmic contents are delivered to the lysosome for degradation. ATG13 localizes on the autophagic isolation membrane and is essential for autophagosome formation. In mammals, ATG13 interacts with ATG1 (ULK1/2) and FIP200 to form the autophagy. ATG101 can also bind to ATG13 protein and is important for the stability and basal phosphorylation of ATG13 and ULK1. mTOR can suppress the autophagy process through direct regulation of the ULK1-Atg13-FIP200 complex where mTOR has been shown to phosphorylate ATG13.

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, 150 mM NaCl, 10 mM glutathione, 0.1 mM EDTA, 0.25 mM DTT, 0.1 mM PMSF, 25% glycerol.

Preparation Note

After opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service