Skip to Content
MilliporeSigma
All Photos(2)

Documents

SRP5324

Sigma-Aldrich

FASTK, active, GST tagged human

PRECISIO®, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

FAST, FLJ13079

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352202
NACRES:
NA.32

biological source

human

recombinant

expressed in baculovirus infected Sf9 cells

product line

PRECISIO®

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

0.8-1.2 nmol/min·mg

mol wt

~84 kDa

NCBI accession no.

application(s)

cell analysis

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... FASTK(10922)

General description

FASTK or Fas-Activated Serine/Threonine Kinase is a member of the serine/threonine protein kinase family that becomes rapidly activated during Fas-mediated apoptosis in Jurkat cells and in response to Fas receptor ligation. FASTK interacts with and phosphorylates TIA1 which is an apoptosis-promoting nuclear RNA-binding protein. FAST K influences alternative pre-mRNA splicing by affecting the activity of TIA-1/TIAR. FASTK is a strong inducer of lymphocyte apoptosis. FASTK is the component of the molecular cascade that involved in FAS-mediated apoptosis. FASTK is highly expreesed in heart, brain, placenta, lung, liver, skeletal muscle, kidney, and pancreas.

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, 150 mM NaCl, 10 mM glutathione, 0.1 mM EDTA, 0.25 mM DTT, 0.1 mM PMSF, 25% glycerol.

Preparation Note

After opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles.

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service