Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SRP5291

Sigma-Aldrich

PLK3, active, GST tagged human

PRECISIO®, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

CNK, FNK, PRK

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
$515.00

$515.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 μG
$515.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

$515.00


Please contact Customer Service for Availability

Request a Bulk Order

recombinant

expressed in baculovirus infected Sf9 cells

product line

PRECISIO®

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

1.8-2.6 nmol/min·mg

mol wt

~60 kDa

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... PLK3(1263)

General description

PLK3 or Polo-like kinase 3 is a cytokine-inducible kinase that is a member of the Polo family of serine/threonine protein kinases. PLK3 contains both a catalytic domain and a putative regulatory domain and plays an important role in the regulation of cell cycle progression and tumorigenesis. PLK3 is closely associated with centrosomes in several human cell lines and its localization is dependent on the integrity of microtubules. Hence, PLK3 is also involved in regulating microtubule dynamics and centrosomal function.

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, 150 mM NaCl, 10 mM glutathione, 0.1 mM EDTA, 0.25 mM DTT, 0.1 mM PMSF, 25% glycerol.

Preparation Note

After opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles.

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service