Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SRP5287

Sigma-Aldrich

SRMS, active, GST tagged human

PRECISIO®, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

C20orf148, SRM, dJ697K14.1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

recombinant

expressed in baculovirus infected Sf9 cells

product line

PRECISIO®

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

23.5-28.8 nmol/min·mg

mol wt

~54 kDa

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... SRMS(6725)

General description

SRMS is a Src-related kinase lacking C-terminal regulatory tyrosine and N-terminal myristylation sites that is involved in proliferation or differentiation of keratinocytes in the skin. SRMS functions as a rapid downstream signaling intermediate following calcium-induced differentiation in keratinocytes. SRMS constitutes a new family of non-receptor tyrosine kinases that may be redundant in function. SRMS can act either in the membrane or at the nucleus, and may change localization patterns depending on external stimuli.

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, 150 mM NaCl, 10 mM glutathione, 0.1 mM EDTA, 0.25 mM DTT, 0.1 mM PMSF, 25% glycerol.

Preparation Note

After opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles.

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service