Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SRP5280

Sigma-Aldrich

PI3K (p110-α/p85-α), active, His tagged from mouse and human

PRECISIO®, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

p110-alpha, p85-alpha

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
$472.00

$472.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 μG
$472.00

About This Item

CAS Number:
UNSPSC Code:
12352200
NACRES:
NA.32

$472.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

human
mouse

recombinant

expressed in baculovirus infected Sf9 cells

product line

PRECISIO®

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

64-97 nmol/min·mg

mol wt

~111 kDa (p110α)
~86 kDa (p85α)

NCBI accession no.

application(s)

cell analysis

shipped in

dry ice

storage temp.

−70°C

Gene Information

mouse ... Pik3ca(18706)

General description

The PI3K comprises of a 110 kDa catalytic subunit and a 85 kDa regulatory subunit. A number of isoforms of the 110 kDa catalytic subunit and the 85 kDa regulatory subunit exist in cells. The p110 alpha catalytic subunit (PIK3CA) is frequently mutated or amplified in a variety of cancers including ovarian and colon. PIK3CA gene copy number is increased in over 30% of ovarian cancers and this leads to increased PI3-kinase activity. Furthermore, the activity of p110 alpha is essential for vascular development and inactivation of p110 alpha leads to severe defects in angiogenic sprouting and vascular remodeling.

Physical form

Supplied in 50 mM sodium phosphate, pH 7.0, 300 mM NaCl, 150 mM imidazole, 0.1 mM PMSF, 0.2 mM DTT, 25% glycerol.

Preparation Note

After opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles.

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service