Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SRP5269

Sigma-Aldrich

HDAC7, active, GST tagged human

recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

DKFZp586J0917, FLJ99588, HD7A, HDAC7A

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

recombinant

expressed in baculovirus infected Sf9 cells

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

64-96 RLU/min·ng

mol wt

~80 kDa

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... HDAC7(51564)

General description

HDAC7 or Histone deacetylase 7 belongs to the histone deacetylase/acuc/apha family and is a component of the histone deacetylase complex. The protein encoded by HDAC7 gene has sequence homology to members of the histone deacetylase family whose protein promotes repression mediated via the transcriptional co-repressor SMRT. HDAC7 interacts with β-catenin keeping endothelial cells in a low proliferation stage. HDAC7 regulates NUR77 and apoptosis in developing thymocytes. HDAC7 is expressed in human osteoarthritis that contributes to cartilage degradation via promoting MMP-13 gene expression.

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, 150 mM NaCl, 10 mM glutathione, 0.1 mM EDTA, 0.25 mM DTT, 0.1 mM PMSF, 25% glycerol.

Preparation Note

After opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service