Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SRP5214

Sigma-Aldrich

ROCK2 (5-554), active, His tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

KIAA0619, ROCK-II, ROKα

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
$582.00

$582.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 μG
$582.00

About This Item

CAS Number:
UNSPSC Code:
12352200
NACRES:
NA.32

$582.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

human

recombinant

expressed in baculovirus infected Sf9 cells

product line

PRECISIO® Kinase

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

514-696 nmol/min·mg

mol wt

~66 kDa

NCBI accession no.

application(s)

cell analysis

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... ROCK2(9475)

General description

ROCK2 is a ubiquitously expressed serine/threonine kinase localized in the nucleus that regulates cytokinesis, smooth muscle contraction, the formation of actin stress fibers and focal adhesions, and the activation of the c-fos serum response element. ROCK2 is an immediate downstream target of the small GTPase RhoA. ROCK2 may play a pivotal role in cardiovascular diseases such as vasospastic angina, ischemic stroke, and heart failure. Inhibition of ROCKs by statins or other selective inhibitors leads to the upregulation and activation of endothelial nitric oxide synthase (eNOS) and reduction of vascular inflammation and atherosclerosis.

Physical form

Supplied in 50mM sodium phosphate, pH 7.0, 300mM NaCl, 150mM imidazole, 0.1mM PMSF, 0.25mM DTT, 25% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kensuke Noma et al.
American journal of physiology. Cell physiology, 290(3), C661-C668 (2006-02-14)
Rho-associated kinases (ROCKs), the immediate downstream targets of RhoA, are ubiquitously expressed serine-threonine protein kinases that are involved in diverse cellular functions, including smooth muscle contraction, actin cytoskeleton organization, cell adhesion and motility, and gene expression. Recent studies have shown
Zhiyong Zhao et al.
Developmental biology, 275(1), 183-191 (2004-10-07)
Development of the endocardial cushions in the heart involves cell migration and cell differentiation, which is known as epithelial-mesenchymal transformation (EMT). These processes are regulated by cell signaling systems. Yet, the roles of intracellular GTPases and their effectors on these

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service