Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP5198

Sigma-Aldrich

KAT3B (532-1153), GST tagged human

recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

p300

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.77

recombinant

expressed in baculovirus infected Sf9 cells

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

mol wt

~120 kDa

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... EP300(2033)

General description

KAT3B (also known as EP300) encodes the adenovirus E1A-associated cellular p300 transcriptional co-activator protein that functions as a histone acetyltransferase. KAT3B regulates transcription via chromatin remodeling and is important in the processes of cell proliferation and differentiation. KAT3B binding is a highly accurate process and can be used for identifying enhancers and their associated activities. KAT3B is also useful in the study of the role of tissue-specific enhancers in human biology and diseases on a genome wide scale. Defects in KAT3B gene are a cause of Rubinstein-Taybi syndrome that play a important role in epithelial cancer and act as a classic tumor suppressor gene.

Physical form

Supplied in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Axel Visel et al.
Nature, 457(7231), 854-858 (2009-02-13)
A major yet unresolved quest in decoding the human genome is the identification of the regulatory sequences that control the spatial and temporal expression of genes. Distant-acting transcriptional enhancers are particularly challenging to uncover because they are scattered among the
S A Gayther et al.
Nature genetics, 24(3), 300-303 (2000-03-04)
The EP300 protein is a histone acetyltransferase that regulates transcription via chromatin remodelling and is important in the processes of cell proliferation and differentiation. EP300 acetylation of TP53 in response to DNA damage regulates its DNA-binding and transcription functions. A

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service