Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP5167

Sigma-Aldrich

BCR (1-695), GST tagged human

recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

ALL, BCR1, CML, D22S11, D22S662, FLJ1645, PHL

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
$342.00

$342.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
20 μG
$342.00

About This Item

CAS Number:
UNSPSC Code:
12352200
NACRES:
NA.32

$342.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

human

recombinant

expressed in baculovirus infected Sf9 cells

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

mol wt

~120 kDa

NCBI accession no.

application(s)

cell analysis

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... BCR(613)

General description

BCR (also known as breakpoint cluster region) encodes a phosphoprotein that has serine/threonine kinase activity. The human c-ABL oncogene is translocated from chromosome 9 to a region on chromosome 22 (associated with BCR) in chronic myelocytic leukemia. In classical t(9;22) translocation, as observed in chronic granulocytic leukemia (CGL), a hybrid DNA unit is produced comprising the BCR gene product plus the translocated c-ABL gene from chromosome 9. The BCR-ABL hybrid protein (p210) is formed that displays increased tyrosine kinase activity. A similar DNA rearrangement of the p210 protein is also found in cases of Philadelphia-positive acute lymphoblastic leukemia (ALL).

Physical form

Supplied in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Eridani et al.
Cytotechnology, 1(1), 33-36 (1987-10-01)
In classical t(9;22) translocation, as observed in chronic granulocytic leukemia (CGL), a hybrid DNA unit is produced, including a rearranged PHL gene, previously known as bcr (breakpoint cluster region) plus the translocated c-abl gene from chromosome 9: a hybrid bcr-abl
K Stam et al.
Molecular and cellular biology, 7(5), 1955-1960 (1987-05-01)
In chronic myelocytic leukemia, the human c-abl oncogene is translocated from chromosome 9 to a region on chromosome 22 designated as the breakpoint cluster region (bcr) (A. de Klein, A. Guerts van Kessel, G. Grosveld, C. R. Bartram, A. Hagemeyer

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service