Skip to Content
MilliporeSigma
All Photos(1)

Documents

SRP5155

Sigma-Aldrich

14-3-3 ζ, GST tagged human

recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

KCIP-1, MGC111427, MGC126532, MGC138156, YWHAZ

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352202
NACRES:
NA.32

biological source

human

recombinant

expressed in baculovirus infected Sf9 cells

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

mol wt

~55 kDa

NCBI accession no.

application(s)

cell analysis

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... YWHAZ(7534)

General description

14-3-3ζ (also known as tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, zeta polypeptide) is a member of the 14-3-3 family of proteins which mediate signal transduction by binding to phosphoserine-containing proteins. 14-3-3ζ protein plays a key role in cancer biology by being an important regulator of major cellular processes such as proliferation, differentiation, senescence and apoptosis. 14-3-3ζ protein has been shown to interact with the IRS1 protein, suggesting a role for this protein in regulating insulin sensitivity by interrupting the association between the insulin receptor and IRS1.

Physical form

Supplied in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zenggang Li et al.
Proceedings of the National Academy of Sciences of the United States of America, 105(1), 162-167 (2007-12-29)
The family of 14-3-3 proteins has emerged as critical regulators of diverse cellular responses under both physiological and pathological conditions. Here, we report an important role of 14-3-3zeta in tumorigenesis through a mechanism that involves anoikis resistance. 14-3-3zeta is up-regulated
M Niemantsverdriet et al.
Oncogene, 27(9), 1315-1319 (2007-08-21)
14-3-3 proteins are relevant to cancer biology as they are key regulators of major cellular processes such as proliferation, differentiation, senescence and apoptosis. So far, the sigma isoform (14-3-3sigma) has most directly been implicated in carcinogenesis and was recognized as

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service