Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP5116

Sigma-Aldrich

PCAF (431-end), GST tagged human

recombinant, expressed in E. coli, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

CAF, GCN5, GCN5L, GCN5L1, P/CAF

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

mol wt

~69 kDa

NCBI accession no.

application(s)

cell analysis

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... KAT2B(8850)

General description

PCAF is a protein that shows homology to the yeast GCN5 and associates with p300/CBP. PCAF has in vitro and in vivo binding activity with CBP and p300, and competes with E1A for binding sites in p300/CBP. PCAF encodes a predicted 832-amino acid protein that showed by RNA blotting to be expressed in all tissues, most strongly in heart and skeletal muscle. PCAF has histone acetyl transferase activity with core histones and nucleosome core particles, indicating that this protein plays a direct role in transcriptional regulation. PCAF is required for MyoD activity and muscle differentiation. PCAF directly acetylated MyoD which then displays an increased affinity for its DNA target.
p300/CBP-associated factor (PCAF) is a member of the histone acetyltransferases (HATs) protein family. It is also known as KAT2B (lysine acetyltransferase 2B), and is the catalytic subunit of the megadalton metazoan complex ATAC (Ada-Two-A containing complex). In animals, this protein is composed of an N terminal extension region, a HAT domain and a bromodomain (BRD). An acetyl-CoA binding pocket is present in the HAT domain, which is globular in structure. This protein is composed of over 2400 amino acids.

Biochem/physiol Actions

p300/CBP-associated factor (PCAF) functions as a co-activator of the tumor suppressor gene p53, and transactivates target genes along with p53, by acetylating both bound p53 and histones within p53 target promoters. Studies in breast cancer cells show that PCAF is a p53 target gene, and its induction results in increased p53 binding to PCAF promoter. This protein interacts with multiple DNA-binding transcriptional factors such as nuclear hormone receptors, CREB (cAMP response element-binding, c-Jun/v-Jun, c-Myb/v-Myb, Sap-1a, c-Fos, and MyoD (myogenic differentiation).

Physical form

Supplied in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dimeric structure of p300/CBP associated factor.
Shi S, et al.
BMC Structural Biology, 14(1), 2-2 (2014)
V Sartorelli et al.
Molecular cell, 4(5), 725-734 (2000-01-05)
p300/CBP and PCAF coactivators have acetyltransferase activities and regulate transcription, cell cycle progression, and differentiation. They are both required for MyoD activity and muscle differentiation. Nevertheless, their roles must be different since the acetyltransferase activity of PCAF but not of
The acetyltransferase p300/CBP-associated factor is a p53 target gene in breast tumor cells.
Watts GS, et al.
Neoplasia, 6(3), 187-194 (2004)
The transcriptional coactivators p300 and CBP are histone acetyltransferases.
Ogryzko VV, et al.
Cell, 87(5), 953-959 (1996)
X J Yang et al.
Nature, 382(6589), 319-324 (1996-07-25)
The adenoviral oncoprotein E1A induces progression through the cell cycle by binding to the products of the p300/CBP and retinoblastoma gene families. A new cellular p300/CBP-associated factor (P/CAF) having intrinsic histone acetylase activity has been identified that competes with E1A.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service