Skip to Content
MilliporeSigma
All Photos(1)

Documents

SRP5113

Sigma-Aldrich

p73a, GST tagged human

recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

TAp73α, TP73

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in baculovirus infected Sf9 cells

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

mol wt

~110 kDa

NCBI accession no.

application(s)

cell analysis

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... TP73(7161)

General description

p73α is a member of the p53 family of transcription factors that are involved in cellular responses to stress and development. P73 has high sequence similarity to p53 and p63, which allows p63 and p73 to transactivate p53-responsive genes causing cell cycle arrest and apoptosis. P73 is a stress-response gene and a downstream effector in the p53 pathway. The p73 protein is expressed at very low levels in normal tissues and is differentially expressed in a number of tumors. p73 is the components of a mismatch repair-dependent apoptosis pathway which contributes to cisplatin-induced cytotoxicity.

Physical form

Supplied in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jianli Wang et al.
The Journal of biological chemistry, 282(40), 29152-29162 (2007-08-19)
TAp73 is a p53 tumor suppressor gene homologue that is known to be mainly involved in apoptosis. We report here that TAp73 is necessary for the cellular response to oxidative stress and that TAp73 functions as a downstream target of
J G Gong et al.
Nature, 399(6738), 806-809 (1999-07-03)
Cancer chemotherapeutic agents such as cisplatin exert their cytotoxic effect by inducing DNA damage and activating programmed cell death (apoptosis). The tumour-suppressor protein p53 is an important activator of apoptosis. Although p53-deficient cancer cells are less responsive to chemotherapy, their

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service