Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SRP5067

Sigma-Aldrich

PKC ε , active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

MGC125656, MGC125657, PRKCE, nPKC-ε

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
$408.00

$408.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 μG
$408.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

$408.00


Please contact Customer Service for Availability

Request a Bulk Order

recombinant

expressed in baculovirus infected Sf9 cells

product line

PRECISIO® Kinase

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

1033-1398 nmol/min·mg

mol wt

~110 kDa

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... PRKCE(5581)

General description

PKCε is a member of the protein kinase C (PKC) family of serine- and threonine-specific protein kinases that can phosphorylate a wide variety of protein targets known to be involved in diverse cellular signaling pathways. PKCε is involved in many different cellular functions, such as neuron channel activation , cardioprotection from ischemia, heat shock response, as well as insulin exocytosis. Knockout studies in mice suggest that this kinase is important for lipopolysaccharide (LPS)-mediated signaling in activated macrophages and may control anxiety-like behavior.

Physical form

Supplied in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Clyde W Hodge et al.
The Journal of clinical investigation, 110(7), 1003-1010 (2002-10-09)
Mice lacking protein kinase Cepsilon (PKCepsilon) are supersensitive to positive allosteric modulators of gamma aminobutyrate type A (GABA(A)) receptors. Since many of these compounds are anxiolytic, we examined whether anxiety-like behavior is altered in these mice. PKCepsilon-null mice showed reduced
C H Chen et al.
Proceedings of the National Academy of Sciences of the United States of America, 96(22), 12784-12789 (1999-10-27)
Recent epidemiological studies indicate beneficial effects of moderate ethanol consumption in ischemic heart disease. Most studies, however, focus on the effect of long-term consumption of ethanol. In this study, we determined whether brief exposure to ethanol immediately before ischemia also

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service