Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SRP5053

Sigma-Aldrich

NEK9 (1-979), (deleted fragment 346-731), active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

DKFZp434D0935, MGC138306, MGC16714, NEK8, NERCC, NERCC1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

recombinant

expressed in baculovirus infected Sf9 cells

product line

PRECISIO® Kinase

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

102-138 nmol/min·mg

mol wt

~115 kDa

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... NEK9(91754)

General description

NEK9 is a member of the NEK family and has high homology to NEK1, NEK3 and NEK4. NEK9 is activated during mitosis and binds specifically to RAN GTPase and is a substrate for CDC2 phosphorylation. NEK9 plays a role in the control of mitotic progression and is regulated by CDC2 and RAN GTPase. Overexpression of both active and inactive variants of NEK9 is toxic to cells and inhibits cell division causing abnormal nuclear morphologies. NEK9 can catalyze the phosphorylation of recombinant NEK6 and NEK7 in vitro leading to its activation. This suggests that NEK9 may be responsible for activation of NEK6 and NEK7 during mitosis.

Physical form

Supplied in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Joan Roig et al.
Genes & development, 16(13), 1640-1658 (2002-07-09)
The protein kinase NIMA is an indispensable pleiotropic regulator of mitotic progression in Aspergillus. Although several mammalian NIMA-like kinases (Neks) are known, none appears to have the broad importance for mitotic regulation attributed to NIMA. Nercc1 is a new NIMA-like
Christopher Belham et al.
The Journal of biological chemistry, 278(37), 34897-34909 (2003-07-04)
The Nek family of protein kinases in humans is composed of 11 members that share an amino-terminal catalytic domain related to NIMA, an Aspergillus kinase involved in the control of several aspects of mitosis, and divergent carboxyl-terminal tails of varying

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service