Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SRP5040

Sigma-Aldrich

IKK α , active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

CHUK, IKBKA, IKK-α, IKK1, NFKBIKA, TCF16

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
$685.00

$685.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 μG
$685.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

$685.00


Please contact Customer Service for Availability

Request a Bulk Order

recombinant

expressed in baculovirus infected Sf9 cells

product line

PRECISIO® Kinase

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

2.1-2.9 nmol/min·mg

mol wt

~114 kDa

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... CHUK(1147)

General description

IKKα is a serine/threonine protein kinase that phosphorylates the I-kappa-B protein which is an inhibitor of the transcription factor NF-kappa-B complex. Phosphorylation of I-kappa-B protein triggers the degradation of the inhibitor via the ubiquitination pathway, thereby activating NF-kappa-B complex. IKKα is an essential regulator of NF-kappa-B-dependent gene expression through control of promoter-associated histone phosphorylation after cytokine exposure. IKKα is a critical component of the cytoplasmic transductional-transcriptional processor leading to induction of IFNα production. IKKα is also involved in the epidermis where it antagonizes mitogenic and angiogenic signals and represses tumor progression and metastases.

Physical form

Supplied in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jiefei Geng et al.
Nature communications, 8(1), 359-359 (2017-08-27)
Stimulation of TNFR1 by TNFα can promote three distinct alternative mechanisms of cell death: necroptosis, RIPK1-independent and -dependent apoptosis. How cells decide which way to die is unclear. Here, we report that TNFα-induced phosphorylation of RIPK1 in the intermediate domain
Vasiliki Anest et al.
Nature, 423(6940), 659-663 (2003-06-06)
NF-kappaB is a principal transcriptional regulator of diverse cytokine-mediated processes and is tightly controlled by the IkappaB kinase complex (IKK-alpha/beta/gamma). IKK-beta and IKK-gamma are critical for cytokine-induced NF-kappaB function, whereas IKK-alpha is thought to be involved in other regulatory pathways.
Katsuaki Hoshino et al.
Nature, 440(7086), 949-953 (2006-04-14)
The Toll-like receptor (TLR) family has important roles in microbial recognition and dendritic cell activation. TLRs 7 and 9 can recognize nucleic acids and trigger signalling cascades that activate plasmacytoid dendritic cells to produce interferon-alpha (IFN-alpha) (refs 7, 8). TLR7/9-mediated

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service