Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SRP5035

Sigma-Aldrich

GRK6, active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

FLJ32135, GPRK6

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

recombinant

expressed in baculovirus infected Sf9 cells

product line

PRECISIO® Kinase

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

9.4-12.6 nmol/min·mg

mol wt

~93 kDa

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... GRK6(2870)

General description

GRK6 is a Ser/Thr protein kinase and a member of the G protein-coupled receptor kinase subfamily. GRK6 is present in all tissues with strongest expression in placenta and skeletal muscle. GRK6 can phosphorylate proteins such as rhodopsin, ADRB2 and the leukotriene B4 receptor BLT1. GRK6 phosphorylates G protein-coupled dopamine receptors, thereby regulating their activity and mediating desensitization of the receptors. GRK6 knockout mice showed supersensitivity to the locomotor-stimulating effects of psychostimulants, including cocaine and amphetamine.

Physical form

Supplied in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Raul R Gainetdinov et al.
Neuron, 38(2), 291-303 (2003-04-30)
Brain dopaminergic transmission is a critical component in numerous vital functions, and its dysfunction is involved in several disorders, including addiction and Parkinson's disease. Responses to dopamine are mediated via G protein-coupled dopamine receptors (D1-D5). Desensitization of G protein-coupled receptors
Remi Gaudreau et al.
The Journal of biological chemistry, 277(35), 31567-31576 (2002-06-22)
Desensitization of G-protein-coupled receptors may involve phosphorylation of serine and threonine residues. The leukotriene B(4) (LTB(4)) receptor (BLT1) contains 14 intracellular serines and threonines, 8 of which are part of consensus target sequences for protein kinase C (PKC) or casein

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service