Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP4695

Sigma-Aldrich

ApoA-1 from rat

recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC)

Synonym(s):

APOA1, Apo-AI, Apolipoprotein A-I, MGC117399

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352202
NACRES:
NA.32

biological source

rat

recombinant

expressed in E. coli

Assay

≥98% (HPLC)
≥98% (SDS-PAGE)

form

lyophilized

mol wt

~27.4 kDa

packaging

pkg of 100 μg

storage condition

avoid repeated freeze/thaw cycles

impurities

endotoxin, tested

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

General description

ApoA-I is a 29.0 kDa protein produced in the liver and intestine, and secreted as the predominant constituent of nascent high-density lipoprotein (HDL) particle. ApoA-I, which is found exclusively in HDL, has a unique ability to capture and solubilize free cholesterol. This apoA-I ability enables HDL to remove excess peripheral cholesterol and return it to the liver for recycling and excretion. This process, called reverse cholesterol transport, is though to inhibit atherogenesis. For this reason HDL is also known as the "good cholesterol." The therapeutic potential of apoA-I has been recently assessed in patients with acute coronary syndromes, using a recombinant form of a naturally occurring variant of apoA-I (called apoA-I Milano). The availability of recombinant normal apoA-I should facilitate further investigation into the potential usefulness of apoA-I in preventing atherosclerotic vascular diseases. Recombinant rat ApoA-I is a 27.4 kDa protein of 235 amino acid residues.

Physical form

Sterile filtered and Lyophilized without additives.

Preparation Note

Centrifuge the vial prior to opening. Avoid freeze-thaw cycles.

Reconstitution

Reconstitute in water to a concentration of 0.1-1.0 mg/ml. The solution can then be diluted into other aqueous buffers and store at 4 °C for 1 week or –20 °C for future use.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service