Skip to Content
MilliporeSigma
All Photos(1)

Documents

SRP4549

Sigma-Aldrich

IL-20 from mouse

recombinant, expressed in E. coli, ≥95% (SDS-PAGE), ≥95% (HPLC)

Synonym(s):

D ZCYTO10, Four α helix cytokine, IL-20, IL10, MGC96907, Zcyto10

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.32

biological source

mouse

recombinant

expressed in E. coli

Assay

≥95% (HPLC)
≥95% (SDS-PAGE)

form

lyophilized

potency

≥100 ng/mL

mol wt

~17.6 kDa

packaging

pkg of 10 μg

storage condition

avoid repeated freeze/thaw cycles

impurities

endotoxin, tested

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

mouse ... Il21(58181)

General description

Mouse IL-20 is fully biologically active when compared to standard. The biological activity was determined by measuring the dose-dependent activation of STAT3 in human epidermal keratinocytes (HEK2a cells). Significant STAT3 activation is observed with >100 ng/ml of recombinant mouse IL-20.

Physical form

Lyophilized without additives.

Reconstitution

Centrifuge the vial prior to opening. Avoid freeze-thaw cycles.
Reconstitute in water to a concentration of 0.1-0.5 mg/mL (Note: Gently pipetting to mix). The solution can be diluted into other aqueous buffers.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service