Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP4542

Sigma-Aldrich

FGF-19 human

recombinant, expressed in E. coli, ≥95% (SDS-PAGE)

Synonym(s):

FGF19, FGFJ, fibroblast growth factor 19

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

Assay

≥95% (SDS-PAGE)

form

lyophilized

potency

50-200 ng/mL

mol wt

~21.8 kDa

packaging

pkg of 25 μg

storage condition

avoid repeated freeze/thaw cycles

impurities

endotoxin, tested

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... FGF19(9965)

General description

Fibroblast growth factor-19 (FGF-19) is an ileum-derived polypeptide hormone and a high affinity heparin-dependent ligand for fibroblast growth factor receptor 4 (FGFR4). It is part of the hormone-like FGF family. The gene encoding FGF-19 is localized on human chromosome 11q13.3. Recombinant human FGF-19 is a 21.8kDa protein containing 195 amino acid residues.

Biochem/physiol Actions

Fibroblast growth factor-19 (FGF-19) is involved in the gut-liver signaling pathway and modulation of bile acid synthesis. Once secreted, it binds to the hepatic fibroblast growth factor receptor 4 (FGFR4)/Klotho-β cell-surface receptor complex and activates it. FGF-19 promotes glycogen synthesis, protein synthesis and lipid metabolism.

Physical form

Lyophilized from 5 mM Tris, pH 7.6 plus 75 mM NaCl. The protein may appear as a haze or film, which you may not see at the bottom of the vial.

Reconstitution

Centrifuge the vial prior to opening. Avoid freeze-thaw cycles.
Reconstitute in water to a concentration of 0.1 to 1.0 μg/μL. This solution can then be diluted into other aqueous buffers.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Identification of a therapeutic strategy targeting amplified FGF19 in liver cancer by Oncogenomic screening.
Sawey ET
Cancer Cell, 19(3), 347-358 (2011)
Targeting fibroblast growth factor 19 in liver disease: a potential biomarker and therapeutic target.
Liu WY
Expert Opinion on Therapeutic Targets, 19(5), 675-685 (2015)
Expression of hepatic Fibroblast Growth Factor 19 is enhanced in Primary Biliary Cirrhosis and correlates with severity of the disease.
Wunsch E
Scientific Reports, 5, 13462-13462 (2015)

Articles

Fibroblast growth factors in cell culture and various growth factors for your research

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service