Skip to Content
MilliporeSigma
All Photos(1)

Documents

SRP4494

Sigma-Aldrich

MIP-3beta human

recombinant, expressed in E. coli, ≥99% (SDS-PAGE), ≥99% (HPLC)

Synonym(s):

CCL19, EBI1-ligand chemokine, ELC, MIP-3-β, Macrophage inflammatory protein 3β, Small inducible cytokine A19

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200

biological source

human

recombinant

expressed in E. coli

Assay

≥99% (HPLC)
≥99% (SDS-PAGE)

form

lyophilized

mol wt

~8.8 kDa

packaging

pkg of 20 μg

impurities

endotoxin, tested

NCBI accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... CCL19(6363)

General description

Human macrophage inflammatory protein-3 beta (MIP-3 beta, Exodus-3, ELC) is a member of the beta-chemokine (C-C) family of cytokines. It plays an important role in the inflammatory response of human T cells and tissue macrophages. Recombinant human MIP-3 beta is a 8.8 kDa protein containing 77 amino acid residues.

Physical form

Lyophilized without any additives.

Reconstitution

Centrifuge the vial prior to opening. Avoid freeze-thaw cycles.
Reconstitute in water to a concentration of 0.1-1.0 mg/mL. This solution can then be diluted into other aqueous buffers.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service