Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP4187

Sigma-Aldrich

IL-27 from mouse

recombinant, expressed in E. coli, ≥97% (SDS-PAGE), ≥97% (HPLC)

Synonym(s):

IL 27, Interleukin27

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

biological source

mouse

recombinant

expressed in E. coli

Assay

≥97% (HPLC)
≥97% (SDS-PAGE)

form

lyophilized

mol wt

~23.7 kDa

packaging

pkg of 10 μg

impurities

endotoxin, tested

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

mouse ... Il27(246779)

General description

Recombinant mouse IL-27/p28 Subunit, also known as Interleukin-30, is a member of the IL-12 family of Cytokines. When combined with EBI3 (Epstein-Barr virus induced gene 3), the heterodimer formed is IL-27. Mouse p28 is a proinflammatory cytokine inducing immunomodulatory effects. Current research is underway to delineate specific biological functions. Recombinant mouse p28 is a single-chain 207 amino acid peptide with a mass of approximately 23.7 kDa.

Physical form

Lyophilized from 10 mM NaP, pH 7.5.

Reconstitution

Centrifuge the vial prior to opening. Avoid freeze-thaw cycles.
Reconstitute in water to a concentration of 0.1-1.0 μg/μL. The solution can then be diluted into other aqueous buffers.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Denada Dibra et al.
PloS one, 6(4), e19072-e19072 (2011-05-12)
Crosstalk between tumor cells and the cognate microenvironment plays a crucial role in tumor initiation and progression. However, only a few genes are known to affect such a crosstalk. This study reveals that WSX1 plays such a role when highly
Juan Liao et al.
PloS one, 9(9), e108777-e108777 (2014-10-01)
The pathogenesis of complex diseases, such as type 1 diabetes (T1D), derives from interactions between host genetics and environmental factors. Previous studies have suggested that viral infection plays a significant role in initiation of T1D in genetically predisposed individuals. T1D
Jui-Hung Yen et al.
Journal of leukocyte biology, 98(5), 689-702 (2015-06-11)
MS is an autoimmune disease characterized by immune cell infiltration in the CNS, leading to cumulative disability. IFN-β, used clinically in RR-MS reduces lesion formation and rates of relapse. Although the molecular mechanisms are not entirely elucidated, myeloid cells appear

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service