Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP4136

Sigma-Aldrich

IL-3 from rat

recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC)

Synonym(s):

HCGF, IL-3, Interleukin-3, MCGF (Mast cell growth factor), MGC79398, Multi-CSF, P-cell stimulation factor

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.32

biological source

rat

recombinant

expressed in E. coli

Assay

≥98% (HPLC)
≥98% (SDS-PAGE)

form

lyophilized

potency

<10 ng/mL ED50

mol wt

~15.8kDa

packaging

pkg of 10 μg

impurities

endotoxin, tested

NCBI accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

rat ... Il3(24495)

General description

Interleukin-3 (IL-3) is a species-specific colony-stimulating factor that stimulates colony formation of megakaryocytes, neutrophils, and macrophages from bone marrow cultures. Recombinant rat IL-3 is a 15.8 kDa protein.

Physical form

Sterile filtered and lyophilized from 5 mM sodium citrate, pH 4.0.

Reconstitution

Centrifuge the vial prior to opening. Avoid freeze-thaw cycles.
Reconstitute in water to a concentration of 0.1-1.0 μg/μL. The solution can be diluted into other aqueous buffers.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Weidong Yu et al.
Carcinogenesis, 29(9), 1717-1724 (2008-05-06)
Abl interactor (Abi) 1 was first identified as the downstream target of Abl tyrosine kinases and was found to be dysregulated in leukemic cells expressing oncogenic Bcr-Abl and v-Abl. Although the accumulating evidence supports a role of Abi1 in actin
Kerstin Kapp et al.
Molecular therapy. Nucleic acids, 3, e170-e170 (2014-06-25)
Toll-like receptors are sensing modulators of the innate immune system. One member of this protein family, Toll-like receptor (TLR)-9, is increasingly being investigated as therapeutic target for infectious diseases and cancer. Double-Stem Loop ImmunoModulator (dSLIM) is a new TLR-9 agonist
Cristina Ghirelli et al.
Cancer research, 75(14), 2775-2787 (2015-05-16)
Reciprocal interactions between tumor cells and their microenvironment vitally impact tumor progression. In this study, we show that GM-CSF produced by primary breast tumor cells induced the activation of plasmacytoid predendritic cells (pDC), a cell type critical to anti-viral immunity.
Orit Uziel et al.
Biochemical and biophysical research communications, 450(1), 274-282 (2014-06-08)
Treatment with erythropoietin (EPO) in several cancers is associated with decreased survival due to cancer progression. Due to the major importance of telomerase in cancer biology we hypothesized that some of these effects may be mediated through EPO effect on
Attila Szabo et al.
Immunology and cell biology, 92(8), 671-678 (2014-05-21)
Plasmacytoid dendritic cells (pDCs) are professional type I interferon (IFN)-producing cells that play an essential role in antiviral immunity. In many cell types, detection of intracellular pathogens is mostly dependent on endosomal Toll-like receptors (TLRs) and cytosolic sensors, such as

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service